Anti-His-Tag Rabbit polyclonal antibody unconjugated
Catalog # GENSA00174-40
Supplier: Genscript
Specifications
- Antibody type:Tag
- Antigen name:His-tag
- Antigen symbol:His
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Specifications
About this item
Type: Tag
Antigen: His-Tag (His-Tag)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: