Anti-CHRNE Goat polyclonal antibody unconjugated
EVBIEB11233
- Antibody type:Primary
- Antigen name:cholinergic receptor, nicotinic, epsilon (muscle)
- Antigen symbol:CHRNE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Amino acid number:185 to 197
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CHRNE (cholinergic receptor, nicotinic, epsilon (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human