Anti-GABRA6 Goat polyclonal antibody unconjugated
EVBIEB10993
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, alpha 6
- Antigen symbol:GABRA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Epitope:N-terminal
- Cross adsorption:none
- Amino acid number:15 to 26
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: GABRA6 (gamma-aminobutyric acid (GABA) A receptor, alpha 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: