Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-PAPD5 Goat Polyclonal Antibody
  EVBIEB10938
undefined
Anti-PAPD5 Goat Polyclonal Antibody
  EVBIEB10938

 

  • Antibody type:
    Primary
  • Antigen name:
    PAP associated domain containing 5
  • Antigen symbol:
    PAPD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Amino acid number:
    497 to 510
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: PAPD5 (PAP associated domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: