Anti-Npc2 (aa21-33) Goat Polyclonal Antibody
EVBIEB10804
- Antibody type:Primary
- Antigen name:Niemann-Pick disease type C2
- Antigen symbol:NPC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Amino acid number:21 to 33
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: Npc2 (aa21-33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat