Anti-Ku70 Rabbit Polyclonal Antibody
BSBTPA1642
- Antibody type:Primary
- Antigen name:5'-deoxyribose-5-phosphate lyase Ku70
- Antigen symbol:Ku70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CTC box binding factor 75 kDa subunit|DNA repair protein XRCC6|lupus Ku autoantigen protein p70|thyroid|Ku autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|70 kDa subunit|ATP-dependent DNA helicase II|5'-dRP lyase Ku70|5'-deoxyribose-5-phosphate lyase Ku70|CTC box-binding factor 75 kDa subunit|70kDa,X-ray repair cross-complementing protein 6|ATP-dependent DNA helicase 2 subunit 1|thyroid autoantigen 70kD (Ku antigen)|ATP-dependent DNA helicase II 70 kDa subunit
- Amino acid number:530 to 545
- Sequence:YPPDYNPEGKVTKRKH
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Ku70 (530-545aa YPPDYNPEGKVTKRKH).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for X-ray repair cross-complementing protein 6(XRCC6) detection. Tested with WB, IHC-P, ICC in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Ku70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human