Specifications
- Antibody type:Primary
- Antigen name:Heat shock protein 90kDa beta member 1
- Antigen symbol:HSP90B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:tumor rejection antigen 1|glucose regulated protein|ECGP|endoplasmin|GP96|GRP-94|94 kDa|heat shock protein 90 kDa beta member 1|Tumor rejection antigen-1|94 kDa glucose-regulated protein|GRP94|TRA1|endothelial cell (HBMEC) glycoprotein|gp96 homolog
- Amino acid number:687 to 700
- UniProtKB:P14625
- Sequence:INPRHPLIRDMLRR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human GRP94 (687-700aa INPRHPLIRDMLRR), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Endoplasmin(HSP90B1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.
Heat shock protein 90kDa beta member 1 (HSP90B1), known also as endoplasmin, GRP94, is a chaperone protein that in humans is encoded by the HSP90B1 gene. It is mapped to chromosome 12q23.3. This gene encodes a member of a family of adenosine triphosphate (ATP)-metabolizing molecular chaperones with roles in stabilizing and folding other proteins. The encoded protein is localized to melanosomes and the endoplasmic reticulum. Expression of this protein is associated with a variety of pathogenic states, including tumor formation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: GRP94
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat