Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-beta-HCG Mouse Monoclonal Antibody [clone: HCG-8]
Anti-beta-HCG Mouse Monoclonal Antibody [clone: HCG-8]
Catalog # BSBTMA1111
Supplier:  Boster Bio
undefined
Anti-beta-HCG Mouse Monoclonal Antibody [clone: HCG-8]
Catalog # BSBTMA1111
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta-HCG
  • Antigen symbol:
    beta-HCG
  • Clonality:
    Monoclonal
  • Clone:
    HCG-8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Purified human Chorionic Gonadotropin.
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Mouse IgG monoclonal antibody for beta-HCG, chorionic gonadotropin, beta polypeptide (CGB) detection. Tested with IHC-P in Human. No cross reactivity with other proteins.

Reconstitute with 1,2% sodium acetate or neutral PBS.

If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.

Type: Primary
Antigen: beta-HCG
Clonality: Monoclonal
Clone: HCG-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human