Anti-Mcm7 Mouse Monoclonal Antibody [clone: M3]
Catalog # BSBTMA1058
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Minichromosome Maintenance Complex Component 7
- Antigen symbol:MCM7
- Clonality:Monoclonal
- Clone:M3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:P85MCM|PNAS146|P1.1-MCM3|P1CDC47|MCM2|CDC47|PPP1R104
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human Mcm7.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Mcm7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human