Anti-VEGFR2 Mouse Monoclonal Antibody [clone: FLK-6]
Catalog # BSBTMA1040
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Kinase insert Domain receptor
- Antigen symbol:KDR
- Clonality:Monoclonal
- Clone:FLK-6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:VEGFR|Kinase insert domain receptor|CD309|VEGFR2|FLK1
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Recombinant human extracellular domain of VEGFR-2 (KDR).
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: VEGFR2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human