Anti-GPR39 Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNLS139G
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 39
- Antigen symbol:GPR39
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:2863
- Antigen synonyms:G-protein coupled receptor 39|GPR39
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µL
- Pk:50 µl
Type: Primary
Antigen: GPR39 (G protein-coupled receptor 39)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse