Mouse IgG2b Isotype Control [clone: MPC-11]
Catalog # NOVUNBP2-27231
Supplier: Novus Biologicals
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MPC-11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: monoclonal
Clone: MPC-11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse