Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: MPC-11] (Alexa Fluor® 647)
Mouse IgG2b Isotype Control [clone: MPC-11] (Alexa Fluor® 647)
Catalog # NOVUNBP2-27228
Supplier:  Novus Biologicals
undefined
Mouse IgG2b Isotype Control [clone: MPC-11] (Alexa Fluor® 647)
Catalog # NOVUNBP2-27228
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MPC-11
  • Conjugation:
    Alexa Fluor® 647
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: monoclonal
Clone: MPC-11
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse