Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-S100A8/A9 Mouse Monoclonal Antibody [clone: 48M7C7]
Anti-S100A8/A9 Mouse Monoclonal Antibody [clone: 48M7C7]
Catalog # NOVUNBP2-25256
Supplier:  Novus Biologicals
undefined
Anti-S100A8/A9 Mouse Monoclonal Antibody [clone: 48M7C7]
Catalog # NOVUNBP2-25256
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 calcium binding protein A8
  • Antigen symbol:
    S100A8
  • Clonality:
    Monoclonal
  • Clone:
    48M7C7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    6279
  • Antigen synonyms:
    L1Ag|CP-10|CFAG|CAGA|MRP8|NIF|MIF|CGLA|60B8AG|MA387|P8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: S100A8/A9
Clonality: monoclonal
Clone: 48M7C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse