Rabbit IgG Isotype Control (FITC (Fluorescein Isothiocyanate))
Catalog # NOVUNBP2-24892
Supplier: Novus Biologicals
Specifications
- Antibody type:Control
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rabbit
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Control
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit