Anti-TET1 Rabbit Polyclonal Antibody
The TET1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TET1. This antibody reacts with mouse. The TET1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: TET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- NOVUNBP1-78965SS
- NOVUNBP1-78965
- Antigen symbol:
- TET1
- TET1
- Antigen name:
- Tet Methylcytosine Dioxygenase 1
- Tet Methylcytosine Dioxygenase 1
- Antigen synonyms:
- CXXC-type zinc finger protein 6|ten-eleven translocation 1 gene protein|bA119F7.1|methylcytosine dioxygenase TET1|CXXC6|tet oncogene 1|leukemia-associated protein with a CXXC domain|ten-eleven translocation-1|LCX|CXXC finger 6|CXXC zinc finger 6
- CXXC-type zinc finger protein 6|ten-eleven translocation 1 gene protein|bA119F7.1|methylcytosine dioxygenase TET1|CXXC6|tet oncogene 1|leukemia-associated protein with a CXXC domain|ten-eleven translocation-1|LCX|CXXC finger 6|CXXC zinc finger 6
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 80312
- 80312
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together