Anti-TGOLN2 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNBP1-49643C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Trans-Golgi Network Integral Membrane Protein 2
- Antigen symbol:TGOLN2
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Gene ID:10618
- Antigen synonyms:TGN38|trans-golgi network protein 2|TGN51|TGN48|TTGN2|TGN46
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
The TGN46 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to TGN46. This antibody reacts with human. The TGN46 Antibody [DyLight 650] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: TGN46
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human