Anti-MAP1LC3A Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # NOVUNBP1-19167PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:microtubule-associated protein 1 light chain 3 alpha
- Antigen symbol:MAP1LC3A
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Gene ID:84557
- Antigen synonyms:MAP1BLC3|MAP1ALC3|LC3A|ATG8E|LC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: