Anti-Oval Cell Marker Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: MIC1-1C3]
NOVUNBP1-18961PE
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:MIC1-1C3
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: MIC1-1C3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse