Anti-HPd1 Mouse Monoclonal Antibody (DyLight® 550) [clone: DHIC2-4A10]
NOVUNBP1-18953R
- Antibody type:Primary
- Antigen symbol:HPD1
- Clonality:Monoclonal
- Clone:DHIC2-4A10
- Conjugation:DyLight® 550
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: HPd1
Clonality: monoclonal
Clone: DHIC2-4A10
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human