Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HPx1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HIC0-3B3]
Anti-HPx1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HIC0-3B3]
Catalog # NOVUNBP1-18951H
Supplier:  Novus Biologicals
Anti-HPx1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: HIC0-3B3]
Catalog # NOVUNBP1-18951H
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HPx1
  • Clonality:
    Monoclonal
  • Clone:
    HIC0-3B3
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human