Anti-HPx1 Mouse Monoclonal Antibody (DyLight® 488) [clone: HIC0-3B3]
NOVUNBP1-18951G
- Antibody type:Primary
- Antigen symbol:HPx1
- Clonality:Monoclonal
- Clone:HIC0-3B3
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: HPx1
Clonality: monoclonal
Clone: HIC0-3B3
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human