Anti-HPi3 Mouse Monoclonal Antibody (DyLight® 350) [clone: HIC1-7H10]
Catalog # NOVUNBP1-18947UV
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:HPI3
- Clonality:Monoclonal
- Clone:HIC1-7H10
- Conjugation:DyLight® 350
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: HPi3
Clonality: monoclonal
Clone: HIC1-7H10
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human