Anti-HPi1 Mouse Monoclonal Antibody (Biotin) [clone: HIC0-4F9]
NOVUNBP1-18872B
- Antibody type:Primary
- Antigen name:Heterochromatin Protein 1
- Antigen symbol:HPI1
- Clonality:Monoclonal
- Clone:HIC0-4F9
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: HPi1
Clonality: monoclonal
Clone: HIC0-4F9
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human