Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307.1]
Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307.1]
Catalog # NOVUNB600-1159
Supplier:  Novus Biologicals
undefined
Anti-PMAIP1 Mouse Monoclonal Antibody [clone: 114C307.1]
Catalog # NOVUNB600-1159
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phorbol-12-myristate-13-acetate-induced protein 1
  • Antigen symbol:
    PMAIP1
  • Clonality:
    Monoclonal
  • Clone:
    114C307.1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    5366
  • Antigen synonyms:
    Immediate-early-response protein APR|PMA-induced protein 1|NOXA|Protein Noxa|Phorbol-12-myristate-13-acetate-induced protein 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

The Noxa Antibody (114C307.1) from Novus Biologicals is a mouse monoclonal antibody to Noxa. This antibody reacts with human, mouse, rat. The Noxa Antibody (114C307.1) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: Noxa
Clonality: Monoclonal
Clone: 114C307.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat