Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD36 Rabbit Polyclonal Antibody (DyLight® 405)
Anti-CD36 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB400-145V
Supplier:  Novus Biologicals
Anti-CD36 Rabbit Polyclonal Antibody (DyLight® 405)
Catalog # NOVUNB400-145V
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD36 molecule (thrombospondin receptor)
  • Antigen symbol:
    CD36
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    948
  • Antigen synonyms:
    GP4|GP3B|FAT|GPIV|SCARB3|CHDS7|BDPLT10|PASIV
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human