Anti-ACSL1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The ACSL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ACSL1. This antibody reacts with human, mouse, rat, primate. The ACSL1 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunocytochemistry / Immunofluorescence.
Simple Western,
Type: Primary
Antigen: ACSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate
Specifications
- Catalog No:
- NOVUNB110-99585
- NOVUNB110-99585SS
- Antigen symbol:
- ACSL1
- ACSL1
- Antigen name:
- Acyl-CoA Synthetase Long-chain Family Member 1
- Acyl-CoA Synthetase Long-chain Family Member 1
- Antigen synonyms:
- FACL1|FACL2|LACS|LACS1|LACS2|ACS1
- FACL1|FACL2|LACS|LACS1|LACS2|ACS1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2180
- 2180
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together