Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-DEFA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8C8]
Anti-DEFA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8C8]
Catalog # NOVUNB110-60002APC
Supplier:  Novus Biologicals
Anti-DEFA5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8C8]
Catalog # NOVUNB110-60002APC
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    defensin, alpha 5, Paneth cell-specific
  • Antigen symbol:
    DEFA5
  • Clonality:
    Monoclonal
  • Clone:
    8C8
  • Conjugation:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    1670
  • Antigen synonyms:
    HD-5|DEF5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: DEFA5 (defensin, alpha 5, Paneth cell-specific)
Clonality: monoclonal
Clone: 8C8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human