Anti-ROBO4 Rabbit Polyclonal Antibody (DyLight 488)
Catalog # NOVUNB110-58778G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Roundabout Homolog 4
- Antigen symbol:ROBO4
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:54538
- Antigen synonyms:MRB|MGC133352|MGC133353|UNQ421/PRO3674|FLJ20798|ECSM4
- Storage buffer:50 mM Sodium Borate
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
The ROBO4 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to ROBO4. This antibody reacts with human. The ROBO4 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml
Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human