Anti-TLR1 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56563
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 1
- Antigen symbol:TLR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:7096
- Antigen synonyms:rsc786|TLR1|Toll-like receptor 1|TIL. LPRS5|KIAA0012|CD281|TIL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TLR1 (toll-like receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat