Anti-Importin-9 Rabbit Polyclonal Antibody
NOVUNB100-56499
- Antibody type:Primary
- Antigen name:importin 9
- Antigen symbol:IPO9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:55705
- Antigen synonyms:Imp9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: Importin-9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: