Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-NTR2 Rabbit Polyclonal Antibody
Anti-NTR2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56472
Supplier:  Novus Biologicals
undefined
Anti-NTR2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56472
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurotensin Receptor 2
  • Antigen symbol:
    NTR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Gene ID:
    23620
  • Antigen synonyms:
    NTR2_HUMAN|Ntsr2.|Levocabastine sensitive neurotensin receptor|NT-R-2|NT R 2|NTR2|Neurotensin receptor type 2|Levocabastine-sensitive neurotensin receptor|NTR2 receptor
  • Storage buffer:
    PBS containing 0,2% gelatin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

The Neurotensin Receptor 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Neurotensin Receptor 2. This antibody reacts with human, mouse. The Neurotensin Receptor 2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 10 ?g/ml

Type: Primary
Antigen: NTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse