Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SEMA3B Rabbit Polyclonal Antibody
Anti-SEMA3B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SEMA3B Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-2218
  • NOVUNB100-2218SS
  • Antigen symbol:
  • SEMA3B
  • SEMA3B
  • Antigen name:
  • Semaphorin 3B
  • Semaphorin 3B
  • Antigen synonyms:
  • SEMA5|SEMAA|LUCA-1|SemA|semaV
  • SEMA5|SEMAA|LUCA-1|SemA|semaV
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 7869
  • 7869
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage buffer:
  • Tris-Glycine and 0,15M NaCl
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together