Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ARNTL Rabbit Polyclonal Antibody
Anti-ARNTL Rabbit Polyclonal Antibody
Catalog # NOVUNB100-129SS
Supplier:  Novus Biologicals
undefined
Anti-ARNTL Rabbit Polyclonal Antibody
Catalog # NOVUNB100-129SS
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ayl hydrocarbon Receptor nuclear translocator-like
  • Antigen symbol:
    BMAL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    406
  • Antigen synonyms:
    ARNTL, BMAL1c, JAP3, MGC47515, MOP3, PASD3, TIC, ARNT-like protein 1, brain and muscle, bHLH-PAS protein JAP3, basic-helix-loop-helix-PAS orphan MOP3, member of PAS superfamily 3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: ARNTL (aryl hydrocarbon receptor nuclear translocator-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human