Anti-SLC30A4 Rabbit Polyclonal Antibody
Catalog # BSBTPA2191
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 30 Member A4
- Antigen symbol:Solute carrier family 30, member 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ZNT4
- Amino acid number:407 - 427
- Sequence:TIQLQSYRQEVDRTCANCQSS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human SLC30A4 (407-427aa TIQLQSYRQEVDRTCANCQSS), different from the related mouse sequence by two amino acids, and from the related rat sequence by three amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Zinc transporter 4(SLC30A4) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: SLC30A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat