Anti-Fibulin 2 Rabbit Polyclonal Antibody
Catalog # BSBTPA2159
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Fibulin 2
- Antigen symbol:FBLN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:OTTHUMP00000216670|Fibulin-2|FIBL-2|Fbln2|OTTHUMP00000216671|FBLN2_HUMAN|OTTHUMP00000216672.
- Amino acid number:188 - 205
- Sequence:DPERHYEDPYSYDQEVAE
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Fibulin 2 (188-205aa DPERHYEDPYSYDQEVAE), different from the related mouse and rat sequences by two amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Fibulin-2(FBLN2) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Fibulin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat