Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-CD134/OX40 Rabbit Polyclonal Antibody
  BSBTPA2136-1
undefined
Anti-CD134/OX40 Rabbit Polyclonal Antibody
  BSBTPA2136-1

 

  • Antibody type:
    Primary
  • Antigen name:
    TNF receptor superfamily member 4
  • Antigen symbol:
    TNFRSF4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    IMD16|TXGP1L|ACT35|OX40|CD134
  • Amino acid number:
    29 - 47
  • Sequence:
    LHCVGDTYPSNDRCCHECR
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human CD134 (29-47aa LHCVGDTYPSNDRCCHECR)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 4(TNFRSF4) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: CD134/OX40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human