Anti-Prothrombin Rabbit Polyclonal Antibody
Catalog # BSBTPA2059
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Coagulation Factor 2
- Antigen symbol:F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:PT|RPRGL2|Prepro-Coagulation Factor II|THPH1|Prothrombin B-Chain
- Amino acid number:600 - 612
- Sequence:THVFRLKRWMQKV
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat Prothrombin (600-612aa THVFRLKRWMQKV)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Prothrombin(F2) detection. Tested with WB in Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Prothrombin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat