Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-NOXA1 Rabbit Polyclonal Antibody
Anti-NOXA1 Rabbit Polyclonal Antibody
  BSBTPA1930
undefined
Anti-NOXA1 Rabbit Polyclonal Antibody
  BSBTPA1930

 

  • Antibody type:
    Primary
  • Antigen name:
    Phorbol-12-myristate-13-acetate-induced protein 1
  • Antigen symbol:
    PMAIP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    Immediate-early-response protein APR|PMA-induced protein 1|NOXA|Protein Noxa|Phorbol-12-myristate-13-acetate-induced protein 1
  • Amino acid number:
    176 - 195
  • Sequence:
    RQVPRGEVFRPHRWHLKHLE
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human NOXA1 (176-195aa RQVPRGEVFRPHRWHLKHLE).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for NADPH oxidase activator 1 (NOXA1) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: NOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human