Anti-E2F6 Rabbit Polyclonal Antibody
BSBTPA1811
- Antibody type:Primary
- Antigen name:E2F transcription Factor 6
- Antigen symbol:E2F6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:MGC111545|E2F6_HUMAN.|E2F binding site modulating activity protein|Transcription factor E2F6|E2F transcription factor 6 isoform 1|EMA|E2F transcription factor 6|E2F 6
- Amino acid number:162 - 177
- Sequence:KDCAQQLFELTDDKEN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human E2F6 (162-177aa KDCAQQLFELTDDKEN), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Transcription factor E2F6 (E2F6) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: E2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat