Anti-ATM Rabbit Polyclonal Antibody
BSBTPA1784
- Antibody type:Primary
- Antigen name:Ataxia Telangiectasia mutated
- Antigen symbol:ATM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:ATM Protein Kinase pS1981
- Amino acid number:36 - 50
- Sequence:DPETIKHLDRHSDSK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human ATM (36-50aa DPETIKHLDRHSDSK), different from the related rat and mouse sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Serine-protein kinase ATM (ATM) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat