Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SNAP23 Rabbit Polyclonal Antibody
Anti-SNAP23 Rabbit Polyclonal Antibody
Catalog # BSBTPA1774
Supplier:  Boster Bio
undefined
Anti-SNAP23 Rabbit Polyclonal Antibody
Catalog # BSBTPA1774
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptosomal-Associated Protein, 23kDa
  • Antigen symbol:
    SNAP23
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    SNAP23A|HsT17016|SNAP23B|SNAP-23
  • Amino acid number:
    192 to 211
  • UniProtKB:
    O00161
  • Sequence:
    DTNRDRIDIANARAKKLIDS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human SNAP23 (192-211aa DTNRDRIDIANARAKKLIDS), different from the related rat and mouse sequences by three amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Synaptosomal-associated protein 23(SNAP23) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

SNAP23(Synaptosomal-Associated Protein, 23-KD), also called SNAP23A, is a protein that in humans is encoded by the SNAP23 gene. The SNAP23 gene has 8 exons, with the initiation codon located in exon 2. The SNAP23 gene is mapped on 15q15.1-q15.2. The SNAP23 cDNA encodes a 211-amino acid polypeptide with a predicted mass of 23 kD. Its amino acid sequence is 59% identical to that of SNAP25. Northern blot analysis revealed that SNAP23 is ubiquitously expressed. SNAP23 is able to bind to multiple syntaxins as well as to multiple vesicle-associated membrane proteins. After relocation, SNAP23 is required for exocytosis, implying a crucial role in promoting membrane fusion. TIVAMP-containing vesicles were concentrated in the apical domain of epithelial cells. STX3A and SNAP23 were codistributed at the apical plasma membrane, where they formed N-ethyl maleimide-dependent SNARE complexes with TIVAMP and cellubrevin. SNAP23 is structurally and functionally similar to SNAP25 and binds tightly to multiple syntaxins and synaptobrevins/VAMPs. It is an essential component of the high affinity receptor for the general membrane fusion machinery and is an important regulator of transport vesicle docking and fusion.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SNAP23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat