Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Gli3 Rabbit Polyclonal Antibody
Anti-Gli3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1766
Supplier:  Boster Bio
undefined
Anti-Gli3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1766
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GLI family zinc finger 3
  • Antigen symbol:
    GLI3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    PAPA|GLI3FL|GCPS|ACLS|PHS|PAPB|GLI3-190|PPDIV|PAP-A|PAPA1
  • Amino acid number:
    40 - 57
  • Sequence:
    SNEDESPGQTYHRERRNA
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human Gli3 (40-57aa SNEDESPGQTYHRERRNA), different from the related rat sequence by two amino acids, and from the related mouse sequence by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Transcriptional activator GLI3 (GLI3) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Gli3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat