Anti-FMO3 Rabbit Polyclonal Antibody
BSBTPA1764
- Antibody type:Primary
- Antigen name:Flavin containing monooxygenase 3
- Antigen symbol:FMO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:dJ127D3.1|FMOII|TMAU
- Amino acid number:76 - 90
- Sequence:DDFPNFMHNSKIQEY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human FMO3 (76-90aa DDFPNFMHNSKIQEY), different from the related rat sequence by one amino acid, and from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Dimethylaniline monooxygenase[N-oxide-forming] 3 (FMO3) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat