Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-MyD88 Rabbit Polyclonal Antibody
Anti-MyD88 Rabbit Polyclonal Antibody
  BSBTPA1660
undefined
Anti-MyD88 Rabbit Polyclonal Antibody
  BSBTPA1660

 

  • Antibody type:
    Primary
  • Antigen name:
    Myeloid differentiation primary response 88
  • Antigen symbol:
    MYD88
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    MYD88D|myeloid differentiation primary response 88
  • Amino acid number:
    174 - 188
  • Sequence:
    FVQEMIRQLEQTNYR
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human MyD88 (174-188aa FVQEMIRQLEQTNYR), different from the related rat and mouse sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Myeloid differentiation primary response protein MyD88(MYD88) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MyD88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat