Specifications
- Antibody type:Primary
- Antigen name:Cytoglobin
- Antigen symbol:CYGB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:STAP|HGB
- Amino acid number:50 to 68
- UniProtKB:Q8WWM9
- Sequence:FVNFPSAKQYFSQFKHMED
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Cytoglobin (50-68aa FVNFPSAKQYFSQFKHMED), identical to the related rat sequence, different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Cytoglobin(CYGB) detection. Tested with WB in Human;Mouse;Rat.
Cytoglobin(CYGB), also called HGB or STAP, is a ubiquitously expressed hexacoordinate hemoglobin that may facilitate diffusion of oxygen through tissues, scavenge nitric oxide or other reactive oxygen species, or serve a protective function during oxidative stress. The cytoglobin gene is mapped on 17q25.1. The CYGB gene contains 4 exons and spans about 9 kb. Cytoglobin has many elements common to vertebrate globins, including invariant histidine residues, and the amino acids that form the heme pocket share similarity with pentacoordinate myoglobin. In contrast to the high oxygen affinities displayed by most hexacoordinate hemoglobins, the characteristics of CYGB indicate that it can facilitate oxygen transport. Because the oxygen affinity of CYGB is more similar to myoglobin than to neuroglobin, and the oxy form of CYGB resists autooxidation, CYGB is proposed to represent a tissue oxygen reservoir by Sawai et al.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Cytoglobin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat