Anti-Kallikrein 1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1633
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein 1
- Antigen symbol:KLK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:243 - 261
- Sequence:YTKLIKFTPWIKEVMKENP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat Kallikrein 1 (243-261aa YTKLIKFTPWIKEVMKENP).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Kallikrein-1(KLK1) detection. Tested with WB, IHC-P in Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Kallikrein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat