Anti-Prostaglandin E Receptor EP1 Rabbit Polyclonal Antibody
BSBTPA1583
- Antibody type:Primary
- Antigen name:Prostaglandin E Receptor EP1
- Antigen symbol:PTGER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:387 - 404
- Sequence:AWEASSLRSSRHSGLSHF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Prostaglandin E Receptor EP1 (387-404aa AWEASSLRSSRHSGLSHF).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Prostaglandin E2 receptor EP1 subtype(PTGER1) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Prostaglandin E Receptor EP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human