Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-TATA binding protein TBP Rabbit Polyclonal Antibody
Anti-TATA binding protein TBP Rabbit Polyclonal Antibody
Catalog # BSBTPA1534
Supplier:  Boster Bio
undefined
Anti-TATA binding protein TBP Rabbit Polyclonal Antibody
Catalog # BSBTPA1534
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TATA-binding protein
  • Antigen symbol:
    TBP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    TATA sequence-binding protein|TFIID|GTF2D|MGC126055|MGC117320|GTF2D1|HDL4|SCA17|RP1-191N21.3|MGC126054|TATA-box factor|transcription initiation factor TFIID TBP subunit|TATA-box binding protein N-terminal domain|TATA-box-binding protein
  • Amino acid number:
    227 to 246
  • UniProtKB:
    P20226
  • Sequence:
    EEQSRLAARKYARVVQKLGF
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human TATA binding protein TBP (227-246aa EEQSRLAARKYARVVQKLGF), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for TATA-box-binding protein(TBP) detection. Tested with WB in Human;Mouse;Rat.

TBP, TATA-binding protein, is a general transcription factor that binds specifically to a DNA sequence called the TATA box. The TBP gene is mapped to 6q27. This DNA sequence is found about 35 base pairs upstream of the transcription start site in some eukaryotic gene promoters. TBP, along with a variety of TBP-associated factors, make up the TFIID, a general transcription factor that in turn makes up part of the RNA polymerase II preinitiation complex. As one of the few proteins in the preinitiation complex that binds DNA in a sequence-specific manner, it helps position RNA polymerase II over the transcription start site of the gene. However, it is estimated that only 10-20% of human promoters have TATA boxes. Therefore, TBP is probably not the only protein involved in positioning RNA polymerase II.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: TATA binding protein TBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat