Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-SOCS2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1383
Supplier:  Boster Bio
undefined
Anti-SOCS2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1383
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Suppressor of Cytokine Signaling 2
  • Antigen symbol:
    SOCS2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    CIS-2|SSI-2|STAT induced STAT inhibitor-2|suppressor of cytokine signaling-2|cytokine-inducible SH2 protein 2|CIS2|STAT-induced STAT inhibitor 2|STAT-induced STAT inhibitor-2|SOCS-2|SSI2|Cish2|STATI2
  • Amino acid number:
    181 to 198
  • UniProtKB:
    O14508
  • Sequence:
    LPLPTRLKDYLEEYKFQV
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human SOCS2 (181-198aa LPLPTRLKDYLEEYKFQV), identical to the related mouse and rat sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Suppressor of cytokine signaling 2(SOCS2) detection. Tested with WB in Human;Mouse;Rat.

Suppressor of cytokine signaling 2 is a protein that in humans is encoded by the SOCS2 gene. This gene encodes a member of the STAT-induced STAT inhibitor(SSI), also known as suppressor of cytokine signalling(SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including erythropoietin, GM-CSF, IL10 and interferon-gamma(IFN-gamma). The protein encoded by this gene is found to interact with the cytoplasmic domain of insulin-like growth factor 1 receptor(IGF1R), and thus is though to be involved in the regulation of IGF1R mediated cell signaling. Knockout studies in mice also suggested a regulatory role of this gene in IGF-1 related growth control. By cytogenetic and radiation hybrid mapping, Yandava et al.(1999) mapped the SOCS2 gene to chromosome 12q21.3-q23.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SOCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat